Cole-Parmer Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, 500 mL

Representative image only.

Cole-Parmer Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, 500 mL

Cole-Parmer | Item: UX-88400-45

Reagents and solutions to meet your lab and research needs

$62.70USD / Each

$62.70USD / Each

Buy It Together

Current Item

Cole-Parmer Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, 500 mL
Cole-Parmer Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, 500 mL
8840045

$62.70USD / Each
+
LabChem Hydrogen Peroxide, 30% (w/w); 500 mL
8002032

$179.25USD / Each
+ =
Total Price 389.70 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeStabilized
  • Empirical FormulaH2O2
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsHydrogen Dioxide, Hyperoxide
  • Volume (mL)500
  • Formula Weight34.01
  • PackagingAmber Poly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7722-84-1, 103-84-4
  • Shelf Life (months)18
  • DescriptionHydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, 500 mL
Reviews
Q&A