LabChem Hydrogen Peroxide, 30% (w/w); 500 mL

Representative image only.

LabChem Hydrogen Peroxide, 30% (w/w); 500 mL

LabChem | Mfr: LC154301| Item: EW-80020-32

Highest quality raw materials!

$179.25USD / Each

$179.25USD / Each

Buy It Together

Current Item

LabChem Hydrogen Peroxide, 30% (w/w); 500 mL
LabChem Hydrogen Peroxide, 30% (w/w); 500 mL
8002032

$179.25USD / Each
+ =
Total Price 241.95 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity Grade
  • Volume (mL)500
  • PackagingPoly Bottle
  • DOT InformationClass 5.1 (8), Packing Group II
  • UN NumberUN2014
  • Shelf Life (days)730
  • Shelf Life (months)24
  • DescriptionHydrogen Peroxide, 30% (w/w); 500 mL
Reviews
Q&A