Cole-Parmer Hydrogen Peroxide, 30% (w/w), ACS Grade, 500 mL

Representative image only.

Cole-Parmer Hydrogen Peroxide, 30% (w/w), ACS Grade, 500 mL

Cole-Parmer | Item: UX-88400-46

Reagents and solutions to meet your lab and research needs

$330.00USD / Each

$330.00USD / Each

Buy It Together

Current Item

Cole-Parmer Hydrogen Peroxide, 30% (w/w), ACS Grade, 500 mL
Cole-Parmer Hydrogen Peroxide, 30% (w/w), ACS Grade, 500 mL
8840046

$330.00USD / Each
+
LabChem Hydrogen Peroxide, 30% (w/w); 500 mL
8002032

$179.25USD / Each
+ =
Total Price 657.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaH2O2
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsHydrogen Dioxide, Hyperoxide
  • Volume (mL)500
  • Formula Weight34.01
  • PackagingVented Poly Bottle
  • DOT InformationHydrogen Peroxide Aqueous Solutions; Class 5.1, Packing Group II
  • CAS Number7722-84-1
  • UN NumberUN2014
  • Shelf Life (months)24
  • DescriptionHydrogen Peroxide, 30% (w/w), ACS Grade, 500 mL
Reviews
Q&A