Cole-Parmer Mixing Bowl, 304 SS, 3/4 Qt Capacity
Representative image only.
Cole-Parmer Mixing Bowl, 304 SS, 3/4 Qt Capacity
Cole-Parmer | Item: EW-07301-00
304 stainless steel composition for increased heat resistance
$15.70USD / Each
BUY MORE & SAVE
  • 1-5$15.70 USD / Each
  • 6-29$14.13 USD / Each
  • 30-119$13.35 USD / Each
  • 120+$12.56 USD / Each
$15.70USD / Each
BUY MORE & SAVE
  • 1-5$15.70 USD / Each
  • 6-29$14.13 USD / Each
  • 30-119$13.35 USD / Each
  • 120+$12.56 USD / Each
Product Description
Key Features
  • Rounded corners for easy mixing and cleaning
  • Withstand temperatures up to 1472 °F (800 °C)
  • 1 year warranty
1 Year WarrantyNSF international certified
Specifications
  • Capacity (L)0.7
  • Capacity (Qt)0.75
  • Material304 stainless steel
  • Overall Height (in)2 3/8
  • Overall Height (cm)6
  • Outside Diameter (OD) (in)6 1/4
  • Outside Diameter (OD) (cm)15.9
  • DescriptionMixing Bowl, 304 SS, 3/4 Qt Capacity; each