Cole-Parmer Mortar and Pestle Set, Porcelain; 125 mL, Each
Representative image only.
Cole-Parmer Mortar and Pestle Set, Porcelain; 125 mL, Each
Cole-Parmer | Item: EW-63100-56
$37.00USD / Each
BUY MORE & SAVE
  • 1-5$37.00 USD / Each
  • 6-29$33.30 USD / Each
  • 30-119$31.45 USD / Each
  • 120+$29.60 USD / Each
$37.00USD / Each
BUY MORE & SAVE
  • 1-5$37.00 USD / Each
  • 6-29$33.30 USD / Each
  • 30-119$31.45 USD / Each
  • 120+$29.60 USD / Each
Buy It Together
Current Item
Cole-Parmer Mortar and Pestle Set, Porcelain; 125 mL, Each
Cole-Parmer Mortar and Pestle Set, Porcelain; 125 mL, Each
6310056
$37.00USD / Each
+
Cole-Parmer 99.5% Alumina Mortar, 7 mL
1782972
$151.75USD / Each
=
Total Price 188.75 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Mortars are glazed on the outside and pestles are glazed to grinding surface. Features pouring spout for easy pouring. Both mortar and pestle are sold as a set.
1 Year Warranty
Specifications
  • Mortars Capacity (mL)125
  • Mortars Top OD (mm)75
  • Mortars Height (mm)50
  • Pestles Length (mm)102
  • DescriptionMortar and Pestle Set, Porcelain; 125 mL, Each