Cole-Parmer Potassium Chloride, ACS Grade, 2500 g

Representative image only.

Cole-Parmer Potassium Chloride, ACS Grade, 2500 g

Cole-Parmer | Item: EW-88404-07

Quality and consistency your lab can count on

$150.00USD / Each

$150.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaKCl
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)2500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7447-40-7
  • Shelf Life (days)60
  • DescriptionPotassium Chloride, ACS Grade, 2500 g
Reviews
Q&A