Cole-Parmer Potassium Permanganate, 0.100N (N/10), 0.0200 Molar (M/50), 4 L

Representative image only.

Cole-Parmer Potassium Permanganate, 0.100N (N/10), 0.0200 Molar (M/50), 4 L

Cole-Parmer | Item: EW-88400-64

Reagents and solutions to meet your lab and research needs

$166.25USD / Each

$166.25USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.1
  • Empirical FormulaKMnO4
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight158.03
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • CAS Number7722-64-7
  • Shelf Life (months)18
  • DescriptionPotassium Permanganate, 0.100N (N/10), 0.0200 Molar (M/50), 4 L
Reviews
Q&A