Cole-Parmer Scalpel Blades, Carbon Steel (CS) #70 Blade; 100/Box
Representative image only.
Cole-Parmer Scalpel Blades, Carbon Steel (CS) #70 Blade; 100/Box
Cole-Parmer | Item: EW-10822-15
$130.25USD / Box of 1
$130.25USD / Box of 1
Buy It Together
Current Item
Cole-Parmer Scalpel Blades, Carbon Steel (CS) #70 Blade; 100/Box
Cole-Parmer Scalpel Blades, Carbon Steel (CS) #70 Blade; 100/Box
1082215
$130.25USD / Box of 1
+ + =
Total Price 215.25 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Ideal for dissection, these blades are available in carbon or stainless steel and individually sealed in foil packs.
Use with dissecting blade handle: 10822-33, 10822-34
Note: Not intended for medical applications or patient use.
Specifications
  • Blade Type70
  • Grade TypeDissecting
  • MaterialCarbon Steel
  • DescriptionScalpel Blades, Carbon Steel (CS) #70 Blade; 100/Box