Cole-Parmer Scalpel Blades, Stainless Steel (SS) #25 Blade; 100/Box
Representative image only.
Cole-Parmer Scalpel Blades, Stainless Steel (SS) #25 Blade; 100/Box
Cole-Parmer | Item: EW-10822-16
$72.90USD / Box of 1
$72.90USD / Box of 1
Buy It Together
Current Item
Cole-Parmer Scalpel Blades, Stainless Steel (SS) #25 Blade; 100/Box
Cole-Parmer Scalpel Blades, Stainless Steel (SS) #25 Blade; 100/Box
1082216
$72.90USD / Box of 1
+ + =
Total Price 157.90 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Ideal for dissection, these blades are available in carbon or stainless steel and individually sealed in foil packs.
Use with dissecting blade handle: 10822-26, 10822-28, 10822-32
Note: Not intended for medical applications or patient use.
Specifications
  • Blade Type25
  • Grade TypeDissecting
  • MaterialStainless Steel
  • DescriptionScalpel Blades, Stainless Steel (SS) #25 Blade; 100/Box