Cole-Parmer Service by AL-TAR On-Site Environmental Chamber IQ/OQ/PQ Qualification
Representative image only.
Cole-Parmer Service by AL-TAR On-Site Environmental Chamber IQ/OQ/PQ Qualification
Cole-Parmer Service by AL-TAR | Mfr: WEB-1031Q| Item: EW-67800-13
No matter where you are, there’s a flat rate for on-site maintenance/calibration, and qualification services, including transportation
$5,480.00USD / Each
$5,480.00USD / Each
Product Description
Key Features
  • Simplify your maintenance and calibration with a partner experienced with multiple types of equipment and major brands
  • AL-TAR’s on-site services for the contiguous 48 states; the price includes travel to your site*
  • Ensure quality service with an ISO 17025 accredited provider
  • You get final say over calibration parameters and tolerances
  • Experienced field engineers with extensive training
  • Document and verify that your new equipment is compatible with your laboratory environment
  • Confirm all utilities and supplies to be within manufacturer specifications
  • Install that equipment according to manufacturer guidelines
  • Evaluate the functionality of your equipment against parameters provided by the manufacturer
  • Test empty chamber/load conditions
  • Verify and document how well your equipment performs throughout all operating ranges with simulated loads
  • Test excursion such as door open and power fail to confirm unit’s ability to recover to original setpoint(s)
More About this Item
Nationwide on-site services are available through our partnership with AL-TAR. Cole-Parmer offers simple service pricing to purchase on-site laboratory equipment services performed by AL-TAR, including travel costs. AL-TAR is an authorized laboratory equipment service representative and provider to over 150 leading manufacturers, dealers, and distributors in the fields of scientific research and biotechnology. Their experienced field engineers have extensive training and technical support resources to draw upon when diagnosing equipment. If replacement is determined to be a better route than repair, the customer experience team at Cole-Parmer is available to help you with your selection.
AL-TAR provides a full-range of services for your laboratory oven. Regardless of the make and model, you can have preventive maintenance and calibration performed or opt for a full IQ/OQ/PQ validation package.
AL-TAR will schedule the on-site services directly with the customer; typically within 2 to 3 business days of an order being placed. Often the services will be reconfirmed a few days ahead of any scheduled service so that everything is in place for a successful visit. During checkout, please enter information in the free-form text box for the specific contact person AL-TAR will work with to coordinate the on-site visit, as well as any other information requested below.
EXISTING EQUIPMENT
When ordering a service for an existing piece of equipment you already have on-site, please include any relevant information about the make and model of the equipment(s) being serviced along with the contact information.
NEW EQUIPMENT
When ordering new equipment along with an on-site service, the scheduling of the service will be planned around the equipment being delivered and installed / ready to use. If you are ordering multiple services for both new and existing equipment, please note that in the checkout text field as services for existing equipment may be able to be scheduled earlier.
Our IQ OQ PQ qualification and validation programs help your company assess, develop and implement regulatory compliance at the level you need, including the FDA’s Rule 21 CFR Part 11 and the EPA’s CROMERRR. As your partner, we can determine the level of compliance and validation appropriate for your company. We customize our services to fit your in-house compliance and qualification / validation programs, allowing you to optimize resources. Our compliance consultants and engineers are experienced in various Quality System aspects, including assessment and evaluation, development and planning, or master plan implementation and review.
As your regulatory compliance partner, we will design a solid strategy for future regulatory compliance objectives as well as provide comprehensive support in meeting instrument validation / qualification and regulatory documentation needs, such as Temperature Mapping and Validation services.
Our qualification / validation offers:
• Complete maintenance history, system history and inventory of components and documentation
• Universal IQ, OQ, PQ template format
• Verification of components’ installation including electrical, plumbing, and communication connections
• Verification of components’ functionality, operation, and conformity to manufacturer’s specifications
• Performance verification of laboratory equipment for routine R&D, QA/QC, EPA, FDA, and client-compliance operation
• Pre-scheduled and annual instrument qualification / validation programs matched to your company’s needs and budget
• Cost effective and fast turnaround for your laboratory equipment IQ, OQ, PQ verification
Flat rate is for services scheduled by our team within a 45-day window from purchase. Expedited service and some exceptions may require additional charges. Bulk rate discount pricing may be available when multiple services are being performed at the same site – call for more information.
Specifications
  • Calibration TypeIQ/OQ/PQ
  • DescriptionEnvironmental Chamber IQ/OQ/PQ Qualification