Cole-Parmer Short Thread Cap, 9mm, Open Red Polypropylene, Red PTFE/Silicone/PTFE Septa; 1000/pk
Representative image only.
Cole-Parmer Short Thread Cap, 9mm, Open Red Polypropylene, Red PTFE/Silicone/PTFE Septa; 1000/pk
Cole-Parmer | Item: EW-98703-45
No more damaged autosampler needles or sample loss
$560.00USD / Pkg of 1000
$560.00USD / Pkg of 1000
Buy It Together
Current Item
Cole-Parmer  Short Thread Cap, 9mm, Open Red Polypropylene, Red PTFE/Silicone/PTFE Septa; 1000/pk
Cole-Parmer Short Thread Cap, 9mm, Open Red Polypropylene, Red PTFE/Silicone/PTFE Septa; 1000/pk
9870345
$560.00USD / Pkg of 1000
+ =
Total Price 607.80 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
  • PTFE/silicone/PTFE septa features PTFE laminated to both sides of high-purity silicone prevents coring
  • Designed with a red open cap with red PTFE laminated white silicone septa
More About this Item
Short thread polypropylene caps fit short thread vials. Caps are available in different colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit using a bonding process without adhesives for contamination-free results. Pre-slit septa prevent vacuum forming inside the vial.
Specifications
  • Cap TypeShort Thread Screw
  • Cap MaterialPolypropylene
  • Cap colorRed
  • Septum TypeRed PTFE/Silicone/PTFE
  • SterileNo
  • Description Short Thread Cap, 9mm, Open Red Polypropylene, Red PTFE/Silicone/PTFE Septa; 1000/pk