Cole-Parmer Sodium Hydroxide, ACS Grade, Pellets, 500 g

Representative image only.

Cole-Parmer Sodium Hydroxide, ACS Grade, Pellets, 500 g

Cole-Parmer | Item: EW-88404-76

Reagents and solutions to meet your lab and research needs

$86.90USD / Each

$86.90USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaNaOH
  • ClassReagent Acids and Bases
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight39.99
  • PackagingPoly Bottle
  • DOT InformationSodium hydroxide, solid; Class 8, Packing Group II
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2
  • UN NumberUN1823
  • Shelf Life (days)60
  • DescriptionSodium Hydroxide, ACS Grade, Pellets, 500 g
Reviews
Q&A