Cole-Parmer Stable Starch Solution, 1 L

Representative image only.

Cole-Parmer Stable Starch Solution, 1 L

Cole-Parmer | Item: EW-88401-56

Quality and consistency your lab can count on

$151.75USD / Each

$151.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.0058
  • Empirical Formula(C6H10O5)_n
  • ClassChemical Indicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number9005-25-8
  • Shelf Life (months)12
  • DescriptionStable Starch Solution, 1 L
Reviews
Q&A