Cole-Parmer Tryptic soy broth, 500g

Representative image only.

Cole-Parmer Tryptic soy broth, 500g

Cole-Parmer | Item: UX-14200-36

Media cultures are tested and standardized to ensure consistent and reliable performance

$151.25USD / Each

$151.25USD / Each
Product Description

Key Features

  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes

More About this Item

A USP soybean-casein digest broth for sterility testing and general-purpose use.

Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service.

Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

Specifications
  • Capacity (g)500
  • DIFCO NameTryptic Soy (0370)
  • BBL NameTrypticase® Soy (11768)
  • DescriptionTryptic Soy Broth, 500 g
Reviews
Q&A