Cole-Parmer® Tryptic Soy Broth
Media cultures are tested and standardized to ensure consistent and reliable performance
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
A USP soybean-casein digest broth for sterility testing and general-purpose use.
Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service.
Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.
$151.25 - $566.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
BBL Name Capacity (g) DIFCO Name
Availability Pricing
COMPARE
Trypticase® Soy (11768) 500 Tryptic Soy (0370)
$151.25USD / Each
COMPARE
Trypticase® Soy (11768) 2000 Tryptic Soy (0370)
$566.00USD / Each