Cole-Parmer Tube Rotator Falcon Tube Holder Disc, 12 Tube Capacity
Representative image only.
Cole-Parmer Tube Rotator Falcon Tube Holder Disc, 12 Tube Capacity
Cole-Parmer | Item: EW-00077-13
Quickly and easily replace tube holder without tools
$263.50USD / Each
$263.50USD / Each
Buy It Together
Current Item
Cole-Parmer Tube Rotator Falcon Tube Holder Disc, 12 Tube Capacity
Cole-Parmer Tube Rotator Falcon Tube Holder Disc, 12 Tube Capacity
0007713
$263.50USD / Each
+ + +
Boekel Scientific 260650 Tube Rotator, 100-240V
5120200
$1,615.00USD / Each
=
Total Price 2555.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Offers an end over end action
  • Holds 12 x 50-mL tubes
  • 1 year warranty
  • Previous part number SB3/3
1 Year Warranty
Specifications
  • Number of Tubes12
  • Tube Size27 to 34 mm dia
  • DescriptionTube Rotator Falcon Tube Holder Disc, 12 tube capacity