Dwyer A-432 Portability Kit for Magnehelic Gauges
Representative image only.
Dwyer A-432 Portability Kit for Magnehelic Gauges
Dwyer | Mfr: A-432| Item: EW-68462-77
$116.50USD / Each
$116.50USD / Each
Buy It Together
Current Item
Dwyer A-432 Portability Kit for Magnehelic Gauges
Dwyer A-432 Portability Kit for Magnehelic Gauges
6846277
$116.50USD / Each
+ =
Total Price 236.50 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Accessories to help your installation and application needs.
Plastic carrying case for Magnehelic® gage of standard range, except high pressure connection, includes 9 ft (2.7m) of 3/16" ID rubber tubing, stand-hang bracket & terminal tube with holder.
1 Year Warranty
Specifications
  • Wetted MaterialsAluminum
  • CaseAluminum
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPortability Kit for Magnehelic Gauges