Environmental Express Analytical Mixture for ICP (23 Components), 100 µg/mL in 4% HNO3 + Tr HF, Solution A; 100 mL

Representative image only.

Environmental Express Analytical Mixture for ICP (23 Components), 100 µg/mL in 4% HNO3 + Tr HF, Solution A; 100 mL

Environmental Express | Item: EW-97014-68

Multi-component analytical standards for ICP analysis

$302.00USD / Each

$302.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
  • Solution A of ICP Analytical Mixture Standard Set 97014-69
  • 100 ppm analytical mixture in 4% HNO3 + Tr HF matrix
  • 100 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 23 components.

• Analytical mixture is a 100 mL bottle which contains 100 µg/mL (100 ppm) of Aluminum, Antimony, Barium, Beryllium, Boron, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Nickel, Potassium, Silicon, Sodium, Strontium, Thallium, Vanadium, Zinc in 4% HNO3 + Tr HF.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • Matrix4% HNO3 & Tr HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAnalytical Mixture for ICP (23 Components), 100 µg/mL in 4% HNO3 & Tr HF, Solution A; 100 mL
Documents & Certificates
Reviews
Q&A