Environmental Express Analytical Mixture Set for ICP (24 Components), 100 µg/mL; 500 mL

Representative image only.

Environmental Express Analytical Mixture Set for ICP (24 Components), 100 µg/mL; 500 mL

Environmental Express | Item: EW-97013-12

Multi-component analytical standards for ICP analysis

$635.00USD / Each

$635.00USD / Each

Buy It Together

Current Item

Environmental Express Analytical Mixture Set for ICP (24 Components), 100 µg/mL; 500 mL
Environmental Express Analytical Mixture Set for ICP (24 Components), 100 µg/mL; 500 mL
9701312

$635.00USD / Each
+ =
Total Price 816.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
  • 100 ppm analytical mixture set contains 2 solutions
  • Two 500 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution set contains 2 solutions.

• Solution A is a 500 mL bottle which contains 100 µg/mL (100 ppm) of Aluminum, Antimony, Barium, Beryllium, Boron, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Nickel, Potassium, Silicon, Sodium, Strontium, Thallium, Vanadium, Zinc in 4% HNO3 + Tr HF. This is available individually as part number 97013-11.

• Solution B is a 500 mL bottle which contains 100 µg/mL (100 ppm) of Silver in 4% HNO3. This is available individually as part number 97013-10.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixMultiple
  • Element/IonTwo bottle set: Solution A and Solution B
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s; Class 8, Packing Group II
  • Hazard SymbolCorrosive, Acute Toxicity
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAnalytical Mixture Set for ICP (24 Components), 100 µg/mL; 500 mL
Reviews
Q&A