Environmental Express APC1620 Clear VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized water; 72/Cs

Representative image only.

Environmental Express APC1620 Clear VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized water; 72/Cs

Environmental Express | Item: EW-35206-57

Minimize hazardous chemical exposure with pre-filled vials

$215.60USD / Case of 72

$215.60USD / Case of 72

Buy It Together

Current Item

Environmental Express APC1620 Clear VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized water; 72/Cs
Environmental Express APC1620 Clear VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized water; 72/Cs
3520657

$215.60USD / Case of 72
+ + =
Total Price 381.95 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
  • Model number APC1620

More About this Item

After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.

Specifications
  • Volume (mL)40
  • Width (InchShort)1 1/8
  • Tube MaterialBorosilicate glass
  • Height (InchShort)3 7/8
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOpen-Top/Septa
  • Cap MaterialPolypropylene
  • Cap colorWhite
  • Thread24 mm
  • SterileNo
  • AutoclavableNo
  • Outside Diameter (in)1 1/8
  • Outside Diameter (mm)27.69
  • Fill5 mL Deionized water
  • Level1
  • Special FeaturesContains stir bar; tare weight listed on label
  • DescriptionClear VOA Vials, 40 mL, Open-Top Cap, Tare Weight, Stir Bar, 5 mL Deionized water; 72/Cs
Reviews
Q&A