Environmental Express Assembled Filter Funnel, PP and MCE, 0.45 um, 50/pk
Representative image only.
Environmental Express Assembled Filter Funnel, PP and MCE, 0.45 um, 50/pk
Environmental Express | Item: EW-35200-39
Convenient units are perfect for vacuum filtration
$115.70USD / Pkg of 1
$115.70USD / Pkg of 1
Buy It Together
Current Item
Environmental Express Assembled Filter Funnel, PP and MCE, 0.45 um, 50/pk
Environmental Express Assembled Filter Funnel, PP and MCE, 0.45 um, 50/pk
3520039
$115.70USD / Pkg of 1
+ + =
Total Price 396.35 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for larger volume filtration
  • Comes standard with pre-inserted filters
More About this Item
These filters fit snugly in most standard vacuum flasks. There are preassembled and disposable options perfect for any number of applications.
Assembled filter funnels with filters include a top cap, filter support and filter.
Specifications
  • Membrane MaterialMixed Cellulose Ester (MCE)
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)25
  • Housing MaterialPolypropylene (PP)
  • Sample Volume (mL)68
  • DescriptionAssembled Filter Funnel, PP and MCE, 0.45 um, 50/pk