Environmental Express BOD Dilution Water Reagent Set, P/Mg/Ca/Fe, 1 L of Each

Representative image only.
Environmental Express Item # EW-53200-31
Easily make BOD dilution water
  • Simple to use—just add 1 mL of each solution to 1 L of water
  • Manufactured according to Standard Methods 5210B for 5-day BOD testing
  • Be in conformance—solutions conform to regulations requiring fresh solutions
  • 3 month warranty

PRODUCT OPTIONS

$170.00USD / Pkg of 4

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagents
  • FormLiquid
  • Volume (mL)4000
  • Packaging4000 mL
  • CAS Number10035-04-8, 10025-77-1, 10034-99-8, 7778-77-0
  • DescriptionBOD Dilution Water Reagent Set; 1000 mL of Each P/Mg/Ca/Fe
  • Warranty3 month
3 Month Warranty

Key Features

  • Simple to use—just add 1 mL of each solution to 1 L of water
  • Manufactured according to Standard Methods 5210B for 5-day BOD testing
  • Be in conformance—solutions conform to regulations requiring fresh solutions
  • 3 month warranty

More About this Item

Reagents and standards are manufactured according to Standard Methods 5210B for 5-day BOD Testing. Solutions conform to regulations regarding fresh solutions. When mixing BOD dilution water add 1 mL of each solution to 1 liter of water.

Kit includes one bottle of 1L for each: Calcium Chloride Solution, 2.75%, Ferric Chloride Solution, 0.025%, Magnesium Sulfate Solution, 2.25%, and Phosphate Buffer Solution, pH 7.2

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat