Environmental Express® BOD Dilution Water Solutions
Easily make BOD dilution water
  • Simple to use—just add 1 mL of each solution to 1 L of water
  • Manufactured according to Standard Methods 5210B for 5-day BOD testing
  • Be in conformance—solutions conform to regulations requiring fresh solutions
Reagents and standards are manufactured according to Standard Methods 5210B for 5-day BOD Testing. Solutions conform to regulations regarding fresh solutions. When mixing BOD dilution water add 1 mL of each solution to 1 liter of water.
$50.30 - $191.50USD / Pkg of 4
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
400 BOD Dilution Water Reagent Set; 100 mL of Each P/Mg/Ca/Fe
$50.30USD / Pkg of 4
COMPARE
4000 BOD Dilution Water Reagent Set; 1000 mL of Each P/Mg/Ca/Fe
$191.50USD / Pkg of 4
COMPARE
2000 BOD Dilution Water Reagent Set; 500 mL of Each P/Mg/Ca/Fe
$121.20USD / Pkg of 4