Environmental Express BOD Dilution Water Reagent Set, P/Mg/Ca/Fe, 500 mL of Each

Representative image only.

Environmental Express BOD Dilution Water Reagent Set, P/Mg/Ca/Fe, 500 mL of Each

Environmental Express | Item: EW-53200-32

Easily make BOD dilution water

$121.20USD / Pkg of 4

$121.20USD / Pkg of 4

Buy It Together

Current Item

Environmental Express BOD Dilution Water Reagent Set, P/Mg/Ca/Fe, 500 mL of Each
Environmental Express BOD Dilution Water Reagent Set, P/Mg/Ca/Fe, 500 mL of Each
5320032

$121.20USD / Pkg of 4
+ =
Total Price 165.50 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Each set includes one bottle each of calcium chloride solution (2.75%), ferric chloride solution (0.025%), magnesium sulfate solution (2.25%), and phosphate buffer solution (pH 7.2).

Key Features

  • Simple to use—just add 1 mL of each solution to 1 L of water
  • Manufactured according to Standard Methods 5210B for 5-day BOD testing
  • Be in conformance—solutions conform to regulations requiring fresh solutions
  • 3 month warranty

More About this Item

Reagents and standards are manufactured according to Standard Methods 5210B for 5-day BOD Testing. Solutions conform to regulations regarding fresh solutions. When mixing BOD dilution water add 1 mL of each solution to 1 liter of water.

Kit includes one bottle of 500mL for each: Calcium Chloride Solution, 2.75%, Ferric Chloride Solution, 0.025%, Magnesium Sulfate Solution, 2.25%, and Phosphate Buffer Solution, pH 7.2

3 Month Warranty
Specifications
  • ClassReagents
  • FormLiquid
  • Volume (mL)2000
  • Packaging2000 mL
  • CAS Number10035-04-8, 10025-77-1, 10034-99-8, 7778-77-0
  • DescriptionBOD Dilution Water Reagent Set; 500 mL of Each P/Mg/Ca/Fe
  • Warranty3 month
Reviews
Q&A