Environmental Express Calcium, 10 µg/mL for ICP-MS in 2% HNO3; 100 mL

Representative image only.

Environmental Express Calcium, 10 µg/mL for ICP-MS in 2% HNO3; 100 mL

Environmental Express | Item: EW-97064-07

Single-component standards for ICP-MS analysis

$54.00USD / Each

$54.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3109
  • For US EPA Methods 200.8 and 6020B
  • 10 ppm calcium in 2% HNO3 matrix
  • 100 mL bottle

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Matrix2% HNO3
  • Element/IonCalcium (Ca)
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionCalcium, 10 µg/mL for ICP-MS in 2% HNO3; 100 mL
Documents & Certificates
Reviews
Q&A