Environmental Express Color-Coded Surrogate, BN/A (100:200); 25 mL

Representative image only.

Environmental Express Color-Coded Surrogate, BN/A (100:200); 25 mL

Environmental Express | Item: EW-35200-71

Prevent costly mistakes by color coding your extraction fluids

$76.15USD / Each

$76.15USD / Each
Product Description

Includes

Certificate of Analysis and SDS.

Key Features

  • Ideal for semi-volatile extractions like EPA Methods 3500C, 625, and 8270
  • Simply add 1 mL of the methanol based indicator to your sample
  • Store in your lab refrigerator

More About this Item

Color coded surrogates are methanol based and supplied in 25mL volumes in screw-top vials — add 1 mL to your sample and help avoid performing re-extractions. These surrogates act as an indicator to ensure the pH range of the sample is correct.

Specifications
  • ClassGeneral Solutions
  • FormLiquid
  • Volume (mL)25
  • Packaging25 mL
  • DOT InformationNot regulated by DOT
  • Storage SymbolRefrigerate
  • Hazard SymbolFlammable, Toxic, Organ damage
  • CAS Number67-56-1, 4165-60-0, 367-12-4, 4165-62-2, 321-60-8, 1718-51-0, and 118-79-6
  • Shelf Life (months)30 (refrigeration required)
  • DescriptionColor-Coded Surrogate, Base Neutral/Acid (100:200); 25 mL
Reviews
Q&A