Environmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk

Representative image only.

Environmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk

Environmental Express | Item: UX-53200-13

Eliminate pipetting and dilutions

$47.00USD / Pkg of 24

$47.00USD / Pkg of 24

Buy It Together

Current Item

Environmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk
Environmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk
5320013

$47.00USD / Pkg of 24
+ =
Total Price 143.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • No more worries about sample contamination
  • Ready-to-use dilutions are easy and convenient
  • For use with 60ml BOD Bottles to acheive a concentration of 198 ppm (mg/L)
  • 3 month warranty

More About this Item

Each vial contains the APHA specified concentration of GGA. Simply shake the vial thoroughly, remove the seal and cap, pour contents, and rinse the vial at least two times with BOD water into the check standard solution. Product has a 24-month shelf life.

GGA Ampulesstable, certified single use ampules for easy GGA test perparations. Twenty-four 1.2 mL vials per package.


Instructions: Shake thoroughly, snap the cap, pour contents, and rinse emptied vial at least 2 times with BOD water into the check standard solution. Each vial contains 1.2mL of the APHA specified concentration of Glucose Glutamic Acid. For use with 60ml BOD Bottles to acheive the method specified concentration of 198 mg/L

3 Month Warranty
Specifications
  • Buffer Kit Values24 ampules
  • Volume (mL)1.2
  • DescriptionEnvironmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk
  • Warranty3 month
Documents & Certificates
Reviews
Q&A