NSI Lab Solutions Environmental Wastewater Snip™ Certified Reference Materials
Validate analytical methods and evaluate the quality of your results with premeasured certified reference materials
  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.
$96.00 - $144.00USD / Pkg of 25
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Description
Availability Pricing
COMPARE
300 mg/L Glucose-Glutamic Acid (GGA) GGA Concentrate Snip™ Certified Reference Material, 300 mg/L, 6 mL; 25/PK
$104.00USD / Pkg of 25
COMPARE
2.50 mg/L Total and Free Chlorine Total Chlorine Accuracy Check Snip™ CRM, 2.50 mg/L, 1.5 mL; 25/PK
$96.00USD / Pkg of 25
COMPARE
0.100 mg/L Total and Free Chlorine Ultra-Low Chlorine Accuracy Check Snip™ CRM, 0.100 mg/L, 1.5 mL; 25/PK
$96.00USD / Pkg of 25
COMPARE
1.00 mg/L Total and Free Chlorine Total Chlorine Accuracy Check Snip™ CRM, 1.00 mg/L, 1.5 mL; 25/PK
$96.00USD / Pkg of 25
COMPARE
0.2% Oil, Grease Oil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK
$144.00USD / Pkg of 20
COMPARE
0.4% Oil, Grease Oil and Grease Snip™ Certified Reference Material, 0.4%, 10 mL; 20/PK
$144.00USD / Pkg of 20