Environmental Express Initial Check Verification (ICV) Standard Set for ICP (28 Components); 100 mL

Representative image only.

Environmental Express Initial Check Verification (ICV) Standard Set for ICP (28 Components); 100 mL

Environmental Express | Item: EW-97014-52

Multi-component ICV standards for ICP analysis

$376.00USD / Each

$376.00USD / Each

Buy It Together

Current Item

Environmental Express Initial Check Verification (ICV) Standard Set for ICP (28 Components); 100 mL
Environmental Express Initial Check Verification (ICV) Standard Set for ICP (28 Components); 100 mL
9701452

$376.00USD / Each
+ =
Total Price 720.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
  • ICV standard set contains 2 solutions
  • Two 100 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard set contains 2 solutions.

• Solution A is a 100 mL bottle which contains 200 µg/mL (200 ppm) of Phosphorus, Potassium, Selenium; 100 µg/mL (100 ppm) of Aluminum, Arsenic, Bismuth, Boron, Calcium, Copper, Iron, Lead, Lithium, Magnesium, Molybdenum, Nickel, Strontium, Thallium; and 50 µg/mL (50 ppm) of Barium, Beryllium, Cadmium, Chromium, Cobalt, Manganese, Vanadium, Zinc in 4% HNO3. This is available individually as part number 97014-51.

• Solution B is a 100 mL bottle which contains 200 µg/mL (200 ppm) of Sulfur; 160 µg/mL (160 ppm) of Sodium; and 100 µg/mL (100 ppm) of Silicon in H2O. This is available individually as part number 97014-50.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonTwo bottle set: Solution A and Solution B
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid, Hydrogen fluoride); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionInitial Check Verification (ICV) Standard Set for ICP (28 Components); 100 mL
Documents & Certificates
Reviews
Q&A