Environmental Express Memory Check Solution Set for ICP-MS (28 Components) in 2% HNO3 + Tr HF; 100 mL

Representative image only.

Environmental Express Memory Check Solution Set for ICP-MS (28 Components) in 2% HNO3 + Tr HF; 100 mL

Environmental Express | Item: EW-97062-54

Multi-component memory check solutions for ICP-MS analysis

$387.00USD / Each

$387.00USD / Each

Buy It Together

Current Item

Environmental Express Memory Check Solution Set for ICP-MS (28 Components) in 2% HNO3 + Tr HF; 100 mL
Environmental Express Memory Check Solution Set for ICP-MS (28 Components) in 2% HNO3 + Tr HF; 100 mL
9706254

$387.00USD / Each
+ =
Total Price 612.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.8 and 6020B
  • Memory check solution set contains 2 solutions
  • Two 100 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution set contains 2 solutions.

• Solution A is a 100 mL bottle which contains 7,200 µg/mL (7,200 ppm) of Chloride; 2,000 µg/mL (2,000 ppm) of Carbon; 1,000 µg/mL (1,000 ppm) of Aluminum, Magnesium, Sodium, Sulfur; and 20 µg/mL (20 ppm) of Antimony, Beryllium, Chromium, Manganese, Nickel, Titanium, Vanadium, Zinc in 2% HNO3 + Tr HF. This is available individually as part number 97062-55.

• Solution B is a 100 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Calcium, Iron, Phosphorus, Potassium; and 20 µg/mL (20 ppm) of Arsenic, Barium, Cadmium, Cobalt, Copper, Lead, Molybdenum, Selenium, Silver, Thallium in 2% HNO3 + Tr HF. This is available individually as part number 97062-56.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix2% HNO3 & Tr HF
  • Element/IonTwo bottle set: Solution A and Solution B
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionMemory Check Solution Set for ICP-MS (28 Components) in 2% HNO3 & Tr HF; 100 mL
Reviews
Q&A