Environmental Express MicroBlock™-R, Block Only, 120 VAC

Representative image only.

Environmental Express MicroBlock™-R, Block Only, 120 VAC

Environmental Express | Item: EW-35209-76

Replacement components for the MicroBlock systems

$9,371.25USD / Each

$9,371.25USD / Each
Product Description

Key Features

  • Reduce chemical waste by 90%
  • Get more accurate distillations
  • Comply with all EPA methodologies for the referenced methods; suitable for cyanide (EPA 335.4), phenol (EPA 420.4) and ammonia (EPA 350.1)
  • 1 year warranty

More About this Item

Minimize heat loss and maintain uniform distillations by using specially designed MicroBlock glassware. Cap distillation tubes after distillation to prevent evaporation or contamination.

1 Year Warranty
Specifications
  • DescriptionMicroBlock™-R, Block Only, 120 VAC
  • Warranty1 year
Reviews
Q&A