Environmental Express ProWeigh® Double Filters, TSS, 76 mm; 100/Pk

Representative image only.

Environmental Express ProWeigh® Double Filters, TSS, 76 mm; 100/Pk

Environmental Express | Item: EW-53200-76

Decrease sample prep time for your suspended solids testing

$229.20USD / Pkg of 100

$229.20USD / Pkg of 100
Product Description

Includes

Each filter is supplied in an aluminum dish that has the weight and filter ID in print and barcode formats.

Key Features

  • Compliant with Standard Methods 2540 Ready-to-use filters for total and volatile suspended solids Each filter is individually prepared, weighed, and labeled to be ready for use upon receipt Eliminate loose fibers and inaccurate weights with precise, laser-cut edges Quality control ensures filter integrity with visual inspection after each cutting, washing, and drying step
  • ProWeigh® Double filters are reweighed after inital weighing and labeling to ensure stable weight and accuracy
  • 3 month warranty

More About this Item

Filters for TSS or VSS.

Filters are composed of binderless borosilicate glass, have 1.5 µm porosity, and are processed thoroughly so you can start testing right out of the box! These TSS filters are vacuum rinsed three times with reagent grade water, dried, cooled, and desiccated to assure the best stability and repeatability.
Filters are robotically weighed and placed in individual aluminum dishes where the weight is then listed in print and barcode format. Our standards demand better than the Standard Methods required tolerance of ±0.5 mg. These filters will reduce labor costs, improve accuracy, and provide quicker reporting times for your lab—all you need to do is filter your sample, dry it, and take the final weight!
Now you can access and download complete data for all ProWeigh Double filters. Data includes box ID, weight 1, weight 2, and lot number.

3 Month Warranty
Specifications
  • MaterialBinderless Borosilicate Glass Microfiber
  • Pore Size (micron)1.5
  • Diameter (mm)76
  • Diameter (cm)7.6
  • DescriptionDouble Weigh Filters, TSS, 76 mm; 100/Pk
  • Warranty3 month
Reviews
Q&A