Environmental Express Selenium +4 Speciation Std, 1,000 µg/mL in H2O; 500 mL

Representative image only.

Environmental Express Selenium +4 Speciation Std, 1,000 µg/mL in H2O; 500 mL

Environmental Express | Item: EW-97061-69

Speciation standards for analyzing species in a sample

$145.00USD / Each

$145.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment. The certificate of analysis includes a lot-specific chromatogram.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3149 for total arsenic and a second source standard for the speciated form
  • 1,000 ppm selenium +4 speciation standard in H2O matrix
  • 500 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixH2O
  • Element/IonSelenium +4
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNot Regulated
  • Hazard SymbolIrritant
  • UN NumberNot Regulated
  • Shelf Life (months)12
  • DescriptionSelenium +4 Speciation Std, 1,000 µg/mL in H2O; 500 mL
Documents & Certificates
Reviews
Q&A