Environmental Express Starter Kit B for ICP (10 Components), 1,000 µg/mL in 2 to 5% HNO3 + Tr HF; 100 mL

Representative image only.

Environmental Express Starter Kit B for ICP (10 Components), 1,000 µg/mL in 2 to 5% HNO3 + Tr HF; 100 mL

Environmental Express | Item: EW-97011-21

Multi-component standards for ICP analysis

$478.00USD / Each

$478.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
  • 1,000 ppm ICP starter kit B in 2 to 5% HNO3 + Tr HFmatrix
  • 100 mL bottle

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 10 components.

• Starter kit B is a 100 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Antimony, Hafnium, Molybdenum, Niobium, Tantalum, Tellurium, Tin, Titanium, Tungsten, and Zirconium in 2 to 5% HNO3 + Tr HF.

A 60 component starter kit which contains 5 solutions (kits A, B, C, D, & E) is available as part number 97011-17.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Matrix2 to 5% HNO3 & Tr HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)18
  • DescriptionStarter Kit B for ICP (10 Components), 1,000 µg/mL in 2 to 5% HNO3 & Tr HF; 100 mL
Documents & Certificates
Reviews
Q&A