Environmental Express Starter Kit for ICP (60 Components), Contains Kits A through E, 1,000 µg/mL; 250 mL

Representative image only.

Environmental Express Starter Kit for ICP (60 Components), Contains Kits A through E, 1,000 µg/mL; 250 mL

Environmental Express | Item: EW-97069-04

Multi-component standards for ICP analysis

$4,089.00USD / Each

$4,089.00USD / Each

Buy It Together

Current Item

Environmental Express Starter Kit for ICP (60 Components), Contains Kits A through E, 1,000 µg/mL; 250 mL
Environmental Express Starter Kit for ICP (60 Components), Contains Kits A through E, 1,000 µg/mL; 250 mL
9706904

$4,089.00USD / Each
+ =
Total Price 5450.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Individual solutions traceable to NIST SRM 3100 series
  • US EPA Methods 200.7 and 6010D
  • 1,000 ppm ICP starter kit contains 5 solutions (kits A, B, C, D, and E)
  • Five 250 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component starter kit contains 5 solutions (kits A, B, C, D, & E).

• Kit A is a 250 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Aluminum, Arsenic, Barium, Beryllium, Bismuth, Cadmium, Calcium, Chromium, Cobalt, Copper, Indium, Iron, Lead, Magnesium, Manganese, Mercury, Nickel, Potassium, Selenium, Silicon*, Silver, Sodium, Strontium, Thallium, Vanadium, Zinc in 2% HNO3. *Silicon from (NH4)2SiF6. This is available individually as part number 97068-99.

• Kit B is a 250 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Antimony, Hafnium, Molybdenum, Niobium, Tantalum, Tellurium, Tin, Titanium, Tungsten, Zirconium in 2 to 5% HNO3 + Tr HF. This is available individually as part number 97069-00.

• Kit C is a 250 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Boron, Phosphorus, Sulfur in H2O. This is available individually as part number 97069-01.

• Kit D is a 250 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Gold, Palladium†, Platinum in 2 to 5% HCl. †Available as Nitrate. This is available individually as part number 97069-02.

• Kit E is a 250 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Cerium, Dysprosium, Erbium, Europium, Gadolinium, Holmium, Lanthanum, Lutetium, Neodymium, Praseodymium, Samarium, Scandium, Terbium, Thorium, Thulium, Uranium, Ytterbium, Yttrium in 2% HNO3. This is available individually as part number 97069-03.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMultiple
  • Element/IonFive bottle kit: Kits A through E
  • FormLiquid
  • Volume (mL)250
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)18
  • DescriptionStarter Kit for ICP (60 Components), Contains Kits A through E, 1,000 µg/mL; 250 mL
Documents & Certificates
Reviews
Q&A