Environmental Express Strontium, 10,000 µg/mL for ICP in 10% HCl; 100 mL

Representative image only.

Environmental Express Strontium, 10,000 µg/mL for ICP in 10% HCl; 100 mL

Environmental Express | Item: EW-97011-94

Single-component standards for ICP analysis

$117.00USD / Each

$117.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3153
  • For US EPA Methods 200.7 and 6010D
  • 10,000 ppm strontium in 10% HCl matrix
  • 100 mL bottle

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10,000 µg/mL (10,000 ppm)
  • Matrix10% HCl
  • Element/IonStrontium (Sr)
  • FormLiquid
  • Volume (mL)100
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Hydrochloric acid); Class 8, Packing Group II
  • Hazard SymbolCorrosive, Irritant
  • UN NumberUN3264
  • Shelf Life (months)18
  • DescriptionStrontium, 10,000 µg/mL for ICP in 10% HCl; 100 mL
Documents & Certificates
Reviews
Q&A