Environmental Express TCLP Extraction Fluid Concentrate, #1; 24 vials
Representative image only.
Environmental Express TCLP Extraction Fluid Concentrate, #1; 24 vials
Environmental Express | Item: UX-35200-15
Save time with these premeasured reagents for EPA Method 1311
$180.20USD / Pkg of 24
$180.20USD / Pkg of 24
Product Description
Includes
Certificate of Analysis and SDS.
Key Features
  • Simply add the vial contents to 2 liters of water
  • Obtain consistent concentration and pH levels from your TCLP fluids
  • 2-year shelf life from the date of production
More About this Item
TCLP extraction fluid concentrates deliver the exact amount of reagents for extraction. Concentrates are supplied in screw-top vials — simply add to 2-liters of laboratory/deionized water and you have Fluid #1 or #2 per EPA Method 1311.