Environmental Express® TCLP Extraction Fluid Concentrates
Save time with these premeasured reagents for EPA Method 1311
  • Simply add the vial contents to 2 liters of water
  • Obtain consistent concentration and pH levels from your TCLP fluids
  • 2-year shelf life from the date of production
TCLP extraction fluid concentrates deliver the exact amount of reagents for extraction. Concentrates are supplied in screw-top vials — simply add to 2-liters of laboratory/deionized water and you have Fluid #1 or #2 per EPA Method 1311.
$114.70 - $180.20USD / Pkg of 24
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
$180.20USD / Pkg of 24
COMPARE
$114.70USD / Pkg of 24