Environmental Express Temperature Probe, with PTFE Holder, Fits 100 mL Digestion Tube
Representative image only.
Environmental Express Temperature Probe, with PTFE Holder, Fits 100 mL Digestion Tube
Environmental Express | Item: EW-53205-79
Temperature probes and timers enhance your hot blocks
$160.95USD / Each
$160.95USD / Each
Product Description
Key Features
  • Temperature probes fit 15, 50, and 100 mL digestion cups
More About this Item
Make digestion easier with these accessories for the Environmental Express HotBlocks.
Specifications
  • Max Temperature (° F)572
  • Max Temperature (° C)300
  • Block TypeDigital Indicator
  • DescriptionTemperature Probe, with PTFE Holder, Fits 100 mL Digestion Tube