Environmental Express Temperature Probe, with PTFE Holder, Fits 50 mL Digestion Tube

Representative image only.

Environmental Express Temperature Probe, with PTFE Holder, Fits 50 mL Digestion Tube

Environmental Express | Item: EW-53205-78

Temperature probes and timers enhance your hot blocks

$90.90USD / Each

$90.90USD / Each
Product Description

Key Features

  • Temperature probes fit 15, 50, and 100 mL digestion cups

More About this Item

Make digestion easier with these accessories for the Environmental Express HotBlocks.

Specifications
  • Max Temperature (° F)572
  • Max Temperature (° C)300
  • Block TypeDigital Indicator
  • DescriptionTemperature Probe, with PTFE Holder, Fits 50 mL Digestion Tube
Reviews
Q&A