Environmental Express Tuning Solution Set for ICP-MS (34 Components); 500 mL

Representative image only.

Environmental Express Tuning Solution Set for ICP-MS (34 Components); 500 mL

Environmental Express | Item: EW-97063-84

Multi-component tuning solutions for ICP-MS analysis

$924.00USD / Each

$924.00USD / Each

Buy It Together

Current Item

Environmental Express Tuning Solution Set for ICP-MS (34 Components); 500 mL
Environmental Express Tuning Solution Set for ICP-MS (34 Components); 500 mL
9706384

$924.00USD / Each
+ =
Total Price 1728.00 ADD 2 ITEMS TO CART
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • For EPA Methods 200.8 and 6020B
  • Tuning solution set contains 2 solutions
  • Two 500 mL bottles

More About this Item

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component tuning solution contains 2 solutions.

• Solution A is a 500 mL bottle which contains 20 µg/mL (20 ppm) of Arsenic, Beryllium, Cadmium, Zinc; 10 µg/mL (10 ppm) of Lead, Magnesium, Nickel; 5 µg/mL (5 ppm) of Aluminum, Barium, Bismuth, Chromium, Cobalt, Copper, Indium, 6Lithium, Lutetium, Manganese, Scandium, Sodium, Strontium, Thallium, Thorium, Uranium, Vanadium; and 2.5 µg/mL (2.5 ppm) of Ytterbium, Yttrium in 5% HNO3. This is available individually as part number 97063-85.

• Solution B is a 500 mL bottle which contains 10 µg/mL (10 ppm) of Antimony, Germanium, Molybdenum, Palladium, Ruthenium, Tin; and 5 µg/mL (5 ppm) of Iridium, Titanium in 10% HCl + 1% HNO3 + Tr HF. This is available individually as part number 97063-86.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonTwo bottle set: Solution A and Solution B
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s. (Nitric acid); Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionTuning Solution Set for ICP-MS (34 Components); 500 mL
Documents & Certificates
Reviews
Q&A