Erlab 483 Resin Work Surface for Vertical Sash Hoods, 49" W

Representative image only.

Erlab 483 Resin Work Surface for Vertical Sash Hoods, 49" W

Erlab | Mfr: SMART483RT| Item: EW-33520-64

$1,100.00USD / Each

$1,100.00USD / Each

Buy It Together

Current Item

Erlab 483 Resin Work Surface for Vertical Sash Hoods, 49" W
Erlab 483 Resin Work Surface for Vertical Sash Hoods, 49" W
3352064

$1,100.00USD / Each
+ =
Total Price 5955.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from Replacement Filters, Work Surfaces, Carts, and Benches

Specifications
  • Hood Width (in)49
  • DescriptionWork Surface for Vertical Sash Hoods, 49" W, resin
Reviews
Q&A