Fisher BioReagents BP2641500 Agar (USP/NF, 80-100 Mesh) (500g) CAS 9002-18-0

Representative image only.

Fisher BioReagents BP2641500 Agar (USP/NF, 80-100 Mesh) (500g) CAS 9002-18-0

Fisher BioReagents | Mfr: BP2641500| Item: EW-88061-32

Quality and consistency your lab can count on

$1,107.00USD / Each

$1,107.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeNF or USP
  • ClassSolids
  • FormPowder
  • Weight (g)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number9002-18-0
  • DescriptionAgar (USP/NF, 80-100 Mesh) (500g) CAS 9002-18-0
Documents & Certificates
Reviews
Q&A