Fisher Chemical Alcohol, Reagent Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Alcohol, Reagent Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: A9624| Item: EW-88001-18

Quality and consistency your lab can count on

$360.00USD / Each

$360.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — Ethanol 89-91% Methanol 4.5-5.5% Isopropyl Alcohol 4.5-5.5%
  • Color (APHA) — ≤10
  • Specific Gravity at 25°C — 0.782-0.793
  • Water — ≤0.10%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableToxicStorage Code Red
Specifications
  • Purity GradeReagent
  • Empirical FormulaCH3CH2OH
  • ClassSolvent Blends
  • ChemistrySolvents
  • FormLiquid
  • SynonymsEthanol, Ethyl Alcohol, Denatured Alcohol, Alcohol, Reagent Alcohol
  • Volume (mL)4000
  • Formula Weight46.07
  • PackagingGlass Bottle
  • DOT InformationAlcohols, N.O.S; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Toxic
  • CAS Number64-17-5 (Ethyl alcohol) 67-56-1 (Methyl alcohol) 67-63-0 (Isopropyl alcohol)
  • UN NumberUN1987
  • Boiling Point C (° C)78.5
  • Melting Point C (° C)-114.1
  • DescriptionAlcohol, Reagent Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A