Fisher Chemical Alcohol, Reagent Grade, Poly Bottle, 1 gal

Representative image only.

Fisher Chemical Alcohol, Reagent Grade, Poly Bottle, 1 gal

Fisher Chemical | Mfr: A962F1GAL| Item: EW-88054-94

Quality and consistency your lab can count on

$299.50USD / Each

$299.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableToxicStorage Code Red
Specifications
  • Purity GradeReagent
  • Empirical FormulaCH3CH2OH
  • ClassSolvent Blends
  • ChemistrySolvents
  • FormLiquid
  • SynonymsEthanol, Ethyl Alcohol, Denatured Alcohol, Alcohol, Reagent Alcohol
  • Volume (mL)3785
  • Formula Weight46.07
  • PackagingPoly Bottle
  • DOT InformationAlcohols, N.O.S; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable, Toxic
  • CAS Number64-17-5 (Ethyl alcohol) 67-56-1 (Methyl alcohol) 67-63-0 (Isopropyl alcohol)
  • UN NumberUN1987
  • Boiling Point C (° C)78.5
  • Melting Point C (° C)-114.1
  • DescriptionAlcohol, Reagent Grade, Poly Bottle, 1 gal
Documents & Certificates
Reviews
Q&A