Fisher Chemical BP1423500 Agar (Granulated/Molecular Genetics) (500g)

Representative image only.

Fisher Chemical BP1423500 Agar (Granulated/Molecular Genetics) (500g)

Fisher BioReagents | Mfr: BP1423500| Item: EW-88057-36

Quality and consistency your lab can count on

$421.75USD / Each

$421.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Storage Code Gray
Specifications
  • Purity GradeMolecular genetics
  • ClassSolids
  • FormPowder
  • Weight (g)500
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number9002-18-0
  • DescriptionAgar (Granulated/Molecular Genetics) (500g)
Documents & Certificates
Reviews
Q&A