Fisher Chemical Calcium Chloride Dihydrate, 100 g

Representative image only.

Fisher Chemical Calcium Chloride Dihydrate, 100 g

Fisher Chemical | Mfr: BP510100| Item: EW-88067-51

Quality and consistency your lab can count on

$131.25USD / Each

$131.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

IrritantStorage Code Gray
Specifications
  • Purity GradeNone
  • Empirical FormulaCaCl2•2H2O
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)100
  • Formula Weight147.01
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number10035-04-8
  • Melting Point C (° C)175
  • DescriptionCalcium Chloride Dihydrate, 100 g
Documents & Certificates
Reviews
Q&A