Fisher Chemical Calcium Chloride Dihydrate, Certified ACS Grade, 500 g

Representative image only.

Fisher Chemical Calcium Chloride Dihydrate, Certified ACS Grade, 500 g

Fisher Chemical | Mfr: C79500| Item: EW-88004-44

Quality and consistency your lab can count on

$239.75USD / Each

$239.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (CaCl2•2H2O ) — 99-105%
  • Insoluble Matter — ≤0.01%
  • pH of a 5% Solution at 25°C — 4.5 to 8.5
  • Oxidizing Substances (as NO3) — ≤0.003%
  • Sulfate — ≤0.01%
  • Ammonium — ≤0.005%
  • Barium — ≤0.005%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤5ppm
  • Magnesium — ≤0.005%
  • Potassium — ≤0.01%
  • Sodium — ≤0.02%
  • Strontium — ≤0.1%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

IrritantStorage Code Gray
Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaCaCl2•2H2O
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)500
  • Formula Weight147.01
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number10035-04-8
  • Melting Point C (° C)175
  • DescriptionCalcium Chloride Dihydrate, Certified ACS Grade, 500 g
Documents & Certificates
Reviews
Q&A