Fisher Chemical Ethyl Alcohol, CDA 19, Histological Grade, 1 gal

Representative image only.

Fisher Chemical Ethyl Alcohol, CDA 19, Histological Grade, 1 gal

Fisher Chemical | Mfr: A406F1GAL| Item: EW-88051-68

Quality and consistency your lab can count on

$364.75USD / Each

$364.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeHistological
  • Empirical FormulaC2H5OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsCompletely Denatured Alcohol, Government Formula 19, CD-19 190 Alcohol
  • Volume (mL)3785
  • Formula Weight46.06
  • PackagingPoly Bottle
  • DOT InformationEthanol Solution; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable
  • UN NumberUN1170
  • CAS Number64-17-5
  • Boiling Point C (° C)79
  • Melting Point C (° C)-85
  • DescriptionEthyl Alcohol, CDA 19, Histological Grade, 1 gal
Reviews
Q&A