Fisher Chemical Ethyl Alcohol, Denatured, Certified Grade, 4 L

Representative image only.

Fisher Chemical Ethyl Alcohol, Denatured, Certified Grade, 4 L

Fisher Chemical | Mfr: A407P4| Item: EW-88051-74

Quality and consistency your lab can count on

$329.75USD / Each

$329.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeCertified
  • Empirical FormulaC2H5OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • SynonymsEthanol denatured, Grain alcohol denatured, Ethyl hydroxide denatured
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight46.06
  • PackagingPoly Bottle
  • DOT InformationEthanol Solution; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable
  • CAS Number64-17-5
  • UN NumberUN1170
  • Boiling Point C (° C)78.5
  • Melting Point C (° C)-90
  • DescriptionEthyl Alcohol, Denatured, Certified Grade, 4 L
Reviews
Q&A